missing translation for 'onlineSavingsMsg'
Learn More

Phospho-Rhodopsin (Ser334) Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marque:  Invitrogen PA538327

Code nomenclature Nacres: NA.41

Informations supplémentaires : Poids : 0.00900kg

Code produit. 15545251

  • 467.00€ / 100µg
Expédition estimée: 04-07-2024
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

The Rho proteins are members of the Ras superfamily of small GTP-binding proteins which have low molecular weight of ∽21 kDa. Studies show that the amino acid sequence of the Rho family is more than 45% identical to each other and 30% or less identical to Ras or Rab/YPT proteins. The Rho proteins play an important role in the organization of actin cytoskeleton, cell adhesion, cell motility and smooth muscle contraction. The Rho proteins are also involved in the signal transduction pathways mediated by the actin cytoskeleton and the activation of the stress activated MAP kinase cascades. Researchers demonstrate that Rho kinase is capable of phosphorylating purified smooth muscle myosin phosphatase and inhibiting its activity in vitro.
Spécification

Spécification

Phospho-Rhodopsin (Ser334)
Polyclonal
Unconjugated
RHO
RHO, CSNBAD1, OPN2, Rhodopsin, RP4, Opsin 2, rod pigment, Opsin-2
Rabbit
Antigen affinity chromatography
RUO
212541, 24717, 6010
-20°C
Liquid
Immunohistochemistry (Paraffin)
1 mg/mL
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4
P08100, P15409, P51489
RHO
A synthetic phosphopeptide derived from human Rhodopsin around the phosphorylation site of Ser334 (D-A-SP-A-T)
100 μg
Primary
Human, Mouse, Rat
Antibody
IgG
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions