Learn More
Phospho-RPS6KA1 (Thr359/Ser363) Rabbit, anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 29225-1-AP-20UL
Informations supplémentaires : Poids : 0.00020kg
Description
The 90 kDa ribosomal S6 kinases (RSK1-4) are a family of widely expressed serine/threonine kinases and play an important role in the MAPK signaling cascade and act downstream of ERK1/2 signaling. It can mediate mitogenic and stress-induced activation of the transcription factors CREB1, ETV1/ER81 and NR4A1/NUR77. RSK1 (RPS6KA1) is also designated as p90RSK associated with cellular proliferation, survival and differentiation. RPS6KA1 has some phosphorylation sites including Ser380, Thr359 and Ser363 which are important for kinase activation (PMID: 16458888, 30813401, 17259979, 10679322)
Spécification
Phospho-RPS6KA1 (Thr359/Ser363) | |
Polyclonal | |
Unconjugated | |
20 μL | |
6195 | |
Liquid |
Western Blot, ELISA | |
200 μg/mL | |
Rabbit | |
Primary | |
Human, Mouse |