missing translation for 'onlineSavingsMsg'
Learn More

PIGZ Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 17384-1-AP-150UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16837264

  • 325.38€ / 150µL
Expédition estimée: 30-09-2022
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

The glycosylphosphatidylinositol (GPI) anchor is a glycolipid found on many blood cells that serves to anchor proteins to the cell surface. This gene encodes a protein that is localized to the endoplasmic reticulum, and is involved in GPI anchor biosynthesis. As shown for the yeast homolog, which is a member of a family of dolichol-phosphate-mannose (Dol-P-Man)-dependent mannosyltransferases, this protein can also add a side-branching fourth mannose to GPI precursors during the assembly of GPI anchors.
Spécification

Spécification

PIGZ
Polyclonal
Unconjugated
PIGZ
GPI mannosyltransferase 4, GPI mannosyltransferase IV, GPI MT IV, hSMP3, PIG Z, PIGZ, SMP3, SMP3 homolog
Rabbit
Antigen Affinity Chromatography
RUO
239827, 303872, 80235
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry
0.23 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q86VD9, Q8BTP0
PIGZ
PIGZ Fusion Protein Ag11259
150 μL
Primary
Rat, Mouse, Human
Antibody
IgG
Vidéos
FDS
Documents
Certificats
Promotions

Promotions