missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PIP Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Marque: Novus Biologicals NBP2-98684-50ul
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
PIP Polyclonal antibody specifically detects PIP in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Spécification
| PIP | |
| Polyclonal | |
| Immunohistochemistry 1:500-1:2000, Immunohistochemistry-Paraffin 1:500-1:2000 | |
| GCDFP15prolactin-inducible protein, GCDFP-15SABP, GPIP4gp17, Gross cystic disease fluid protein 15, prolactin-induced proteinSecretory actin-binding protein | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human PIP. | |
| 50 μg | |
| Cancer, Cellular Markers, Signal Transduction | |
| 5304 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu