Learn More
PIP5K1A Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque:  Proteintech 15713-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Code produit. 16835014
  • 140.00€ / 20µL
Expédition estimée: 14-05-2024
Description
The PIP5K1A antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human PIP5K1A. This antibody recognizes human, mouse, rat antigen. The PIP5K1A antibody has been validated for the following applications: FC, IF, IHC, IP, WB, ELISA analysis.
Spécification
Spécification
PIP5K1A
Polyclonal
Unconjugated
20 μL
8394
IgG
Western Blot, Immunoprecipitation, Immunohistochemistry, Immunofluorescence, Flow Cytometry
400 μg/mL
Rabbit
Primary
Human, Rat, Mouse