Learn More
PIP5K1A Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 15713-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
The PIP5K1A antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human PIP5K1A. This antibody recognizes human, mouse, rat antigen. The PIP5K1A antibody has been validated for the following applications: FC, IF, IHC, IP, WB, ELISA analysis.
Spécification
PIP5K1A | |
Polyclonal | |
Unconjugated | |
20 μL | |
8394 | |
IgG |
Western Blot, Immunoprecipitation, Immunohistochemistry, Immunofluorescence, Flow Cytometry | |
400 μg/mL | |
Rabbit | |
Primary | |
Human, Rat, Mouse |