Learn More
PIRT Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque:  Proteintech 20990-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Code produit. 16830585
  • 121.38€ / 20µL
Expédition estimée: 28-09-2022
Description
PIRT is a key component of the TRPV1 complex that positively regulates TRPV1 activity. It binds various phosphoinositide, including phosphatidylinositol 4, 5-bisphosphate (PIP2), but not phosphatidylinositol (PI). PIRT is a membrane protein with MW 15-25 kDa.
Spécification
Spécification
PIRT
Polyclonal
Unconjugated
PIRT
hCG_1776018, PIRT, TRPV1 complex
Rabbit
Antigen Affinity Chromatography
RUO
644139
-20°C
Liquid
Western Blot
0.45 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P0C851
PIRT
Peptide