missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PKN2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 14608-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
PKC-related serine/threonine-protein kinase and Rho/Rac effector protein that participates in specific signal transduction responses in the cell. Plays a role in the regulation of cell cycle progression, actin cytoskeleton assembly, cell migration, cell adhesion, tumor cell invasion and transcription activation signaling processes. Phosphorylates CTTN in hyaluro-induced astrocytes and hence decreases CTTN ability to associate with filamentous actin. Phosphorylates HDAC5, therefore lead to impair HDAC5 import. Direct RhoA target required for the regulation of the maturation of primordial junctions into apical junction formation in bronchial epithelial cells. Required for G2/M phases of the cell cycle progression and abscission during cytokinesis in a ECT2-dependent manner. Stimulates FYN kinase activity that is required for establishment of skin cell-cell adhesion during keratinocytes differentiation. Regulates epithelial bladder cells speed and direction of movement during cell migration and tumor cell invasion. Inhibits Akt pro-survival-induced kinase activity. Mediates Rho protein-induced transcriptional activation via the c-fos serum response factor (SRF). Phosphorylates HCV NS5B leading to stimulation of HCV RNA replication.Spécification
PKN2 | |
Polyclonal | |
Unconjugated | |
PKN2 | |
Pak 2, PAK2, PKN2, PRK2, PRKCL2, PRO2042, Protein kinase C like 2, protein kinase N2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
109333, 207122, 5586 | |
-20°C | |
Liquid |
Immunoprecipitation, Flow Cytometry, Western Blot, Immunocytochemistry, Immunofluorescence | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O08874, Q16513, Q8BWW9 | |
PKN2 | |
PKN2 Fusion Protein Ag6164 | |
20 μL | |
Primary | |
Rat, Human, Mouse | |
Antibody | |
IgG |