missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PKNOX1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 10614-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
PKNOX1 gene ontology annotations related to this gene include regulation of transcription, DNA-templated.Spécification
PKNOX1 | |
Polyclonal | |
Unconjugated | |
PKNOX1 | |
Homeobox protein PKNOX1, Homeobox protein PREP 1, PBX/knotted 1 homeobox 1, PBX/knotted homeobox 1, PKNOX1, pkonx1c, PREP1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
18771, 294322, 5316 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O70477, P55347 | |
PKNOX1 | |
PKNOX1 Fusion Protein Ag0939 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |