missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Placental lactogen Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 16326-1-AP-150UL
Code nomenclature Nacres: NA.41
Informations supplémentaires : Poids : 0.00010kg
Description
The protein encoded by this gene is a member of the somatotropin/prolactin family of hormones and plays an important role in growth control. The gene is located at the growth hormone locus on chromosome 17 along with four other related genes in the same transcriptional orientation; an arrangement which is thought to have evolved by a series of gene duplications. Although the five genes share a remarkably high degree of sequence identity, they are expressed selectively in different tissues. Alternative splicing generates additional isoforms of each of the five growth hormones. This particular family member is expressed mainly in the placenta and utilizes multiple transcription initiation sites. Expression of the identical mature proteins for chorionic somatomammotropin hormones 1 and 2 is upregulated during development, while the ratio of 1 to 2 increases by term. Structural and expression differences provide avenues for developmental regulation and tissue specificity.Spécification
Placental lactogen | |
Polyclonal | |
Unconjugated | |
CSH2 | |
Choriomammotropin, CS 2, CSB, CSH1, CSH2, hCS B, Lactogen, PL, Placental lactogen | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
1443 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.17 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P0DML3 | |
CSH2 | |
Placental lactogen Fusion Protein Ag9412 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |