missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLCB3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 21370-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
PLCB3 encodes a member of the phosphoinositide phospholipase C beta enzyme family that catalyze the production of the secondary messengers diacylglycerol and inositol 1,4,5-triphosphate from phosphatidylinositol in G-protein-linked receptor-mediated signal transduction. Alternative splicing results in multiple transcript variants.Spécification
PLCB3 | |
Polyclonal | |
Unconjugated | |
PLCB3 | |
Phospholipase C beta 3, PLC beta 3, PLCB3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
18797, 5331 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.37 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P51432, Q01970 | |
Plcb3 | |
PLCB3 Fusion Protein Ag15845 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |