missing translation for 'onlineSavingsMsg'
Learn More
Learn More
POLR2K Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Marque: Novus Biologicals NBP2-93400-0.1ml
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
POLR2K Polyclonal antibody specifically detects POLR2K in Human, Mouse, Rat samples. It is validated for Western Blot
Spécification
| POLR2K | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| ABC10-alpha, DNA directed RNA polymerases I, II, and III 7.0 kda polypeptide, DNA-directed RNA polymerase II subunit K, DNA-directed RNA polymerases I, II, and III subunit RPABC4, hRPB7.0, hsRPB10a, polymerase (RNA) II (DNA directed) polypeptide K, 7.0kDa, RNA polymerase II 7.0 kDa subunit, RNA polymerases I, II, and III subunit ABC4, RPABC4, RPB10alphapolymerase (RNA) II (DNA directed) polypeptide K (7.0kD), RPB12, RPB7.0 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-58 of human POLR2K (NP_005025.1). MDTQKDVQPPKQQPMIYICGECHTENEIKSRDPIRCRECGYRIMYKKRTKRLVVFDAR | |
| 0.1 mL | |
| DNA Repair, DNA replication Transcription Translation and Splicing | |
| 5440 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu