missing translation for 'onlineSavingsMsg'
Learn More
Learn More
POLR2L Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 15779-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
This gene encodes a subunit of RNA polymerase II, the polymerase responsible for synthesizing messenger RNA in eukaryotes. The product of this gene contains four conserved cysteines characteristic of an atypical zinc-binding domain. Like its counterpart in yeast, this subunit may be shared by the other two DNA-directed RNA polymerases.Spécification
POLR2L | |
Polyclonal | |
Unconjugated | |
POLR2L | |
hRPB7.6, hsRPB10b, POLR2L, RBP10, RPABC5, RPB10, RPB10 homolog, RPB10beta, RPB7.6 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
502374, 5441, 66491 | |
-20°C | |
Liquid |
Western Blot | |
0.29 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P62875, P62876 | |
POLR2L | |
POLR2L Fusion Protein Ag8468 | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG |