missing translation for 'onlineSavingsMsg'
Learn More

POTED Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 20206-1-AP-150UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16829724

  • 325.38€ / 150µL
Expédition estimée: 28-09-2022
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

The antibody is specific to POTED.

The POTE protein contains seven ankyrin repeats between amino acids 140 and 380. Expression of POTE in prostate cancer and its undetectable expression in normal essential tissues make POTE a candidate for the immunotherapy of prostate cancer. The existence of a large number of closely related but rapidly diverging members, their location on multiple chromosomes and their limited expression pattern suggest an important role for the POTE gene family in reproductive processes.
Spécification

Spécification

POTED
Polyclonal
Unconjugated
POTED
A26B3, ANKRD21, ANKRD26 like family B member 3, POTE, POTE 21, POTE21, POTED, Protein POTE
Rabbit
Antigen Affinity Chromatography
RUO
317754
-20°C
Liquid
Western Blot
0.17 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q86YR6
POTED
Peptide
150 μL
Primary
Mouse, Human
Antibody
IgG
Vidéos
FDS
Documents
Certificats
Promotions

Promotions