missing translation for 'onlineSavingsMsg'
Learn More

PPAP2A Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 17967-1-AP-20UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16837984

  • 121.38€ / 20µL
Expédition estimée: 30-09-2022
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Phosphatidic acid phosphatase type 2 (PAP2) was originally identified as a plasma membrane enzyme that catalyses the dephosphorylation of the putative second messenger, phosphatidic acid (PA) to diacylglycerol (DG). Subsequently, multiple isoforms of PAP2 were cloned [2-5]. It was found that these enzymes dephosphorylate a number of lipid phosphates in vitro other than PA, including the potent bioactive lipids, lysophosphatidic acid (LPA) and sphingosine 1-phosphate (S1P). Therefore, they have been renamed lipid phosphate phosphatases (LPPs). Currently, there are four members of this family called LPP1, LPP1a, LPP2 and LPP3.
Spécification

Spécification

PPAP2A
Polyclonal
Unconjugated
PLPP1
LLP1a, LPP1, PAP 2a, PAP2, PAP2 alpha, PAP2a, PAP2a2, PAP2alpha2, PAPalpha1, PPAP2A
Rabbit
Antigen Affinity Chromatography
RUO
8611
-20°C
Liquid
Western Blot, Immunoprecipitation
0.21 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O14494
PLPP1
PPAP2A Fusion Protein Ag11615
20 μL
Primary
Human
Antibody
IgG
Vidéos
FDS
Documents
Certificats
Promotions

Promotions