missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPAR gamma Mouse anti-Human, Clone: 4E12F10, Proteintech
Mouse Monoclonal Antibody
Marque: Proteintech 60127-1-IG-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
PPP1R10 is a protein with similarity to a rat protein that has an inhibitory effect on protein phosphatase-1 (PP1). The rat protein localizes to the nucleus and colocalizes with chromatin at distinct phases during mitosis. This gene encodes a protein with similarity to a rat protein that has an inhibitory effect on protein phosphatase-1 (PP1). The rat protein localizes to the nucleus and colocalizes with chromatin at distinct phases during mitosis. This gene lies within the major histocompatibility complex class I region on chromosome 6.Spécification
PPAR gamma | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P37231 | |
PPARG | |
PPAR gamma Fusion Protein Ag10005 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA, Flow Cytometry, Western Blot | |
4E12F10 | |
Unconjugated | |
PPARG | |
CIMT1, NR1C3, PPAR gamma, PPARG, PPARG1 | |
Mouse | |
Caprylic Acid Precipitation | |
RUO | |
5468 | |
-20°C | |
Liquid |