Learn More
PPAR gamma Polyclonal Antibody, Invitrogen™
Chicken Polyclonal Antibody
Marque: Invitrogen PA19576
Code nomenclature Nacres: NA.41
Informations supplémentaires : Poids : 0.01000kg
Description
PA1-9576 detects PPARg in human samples. PA1-9576 has been successfully used in Western blot procedures. By Western blot, PA1-9576 detects a 58 kDa protein representing PPARg. PA1-9576 immunogen is a synthetic peptide based on human PPARg protein
This gene encodes a member of the peroxisome proliferator-activated receptor (PPAR) subfamily of nuclear receptors. PPARs form heterodimers with retinoid X receptors (RXRs) and these heterodimers regulate transcription of various genes. Three subtypes of PPARs are known: PPAR-alpha, PPAR-delta, and PPAR-gamma. The protein encoded by this gene is PPAR-gamma and is a regulator of adipocyte differentiation. Additionally, PPAR-gamma has been implicated in the pathology of numerous diseases including obesity, diabetes, atherosclerosis and cancer. Alternatively spliced transcript variants that encode different isoforms have been described.Spécification
PPAR gamma | |
Polyclonal | |
Unconjugated | |
PPARG | |
CIMT1; GLM1; HUMPPARG; NR1C3; nuclear hormone receptor; nuclear hormone receptor; PPARG; Nuclear receptor subfamily 1 group C member 3; nuclear receptor transcription factor; OTTHUMP00000185032; OTTHUMP00000185036; peroxisome proliferative activated receptor, gamma; peroxisome proliferative activated receptor, gamma-2; peroxisome proliferator; peroxisome proliferator activated receptor; peroxisome proliferator activated receptor gamma; peroxisome proliferator activated receptor gamma 1; peroxisome proliferator activated receptor gamma 2; peroxisome proliferator activated receptor gamma 4; peroxisome proliferator activated receptor gamma-1; peroxisome proliferator activated receptor gamma-2; peroxisome proliferator activatived receptor gamma; peroxisome proliferator activator receptor, gamma; peroxisome proliferator-activated nuclear receptor gamma variant 1; peroxisome proliferator-activated receptor; peroxisome proliferator-activated receptor gamma; peroxisome proliferator-activated r | |
Chicken | |
Affinity chromatography | |
RUO | |
5468 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry, Western Blot | |
0.5 mg/mL | |
PBS with 0.02% sodium azide | |
P37231 | |
PPARG | |
Human PPARg peptide AA87-101 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgY |