Learn More
PREX2 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque:  Proteintech 24195-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
 Afficher plus de versions de ce produit
Code produit. 16832715
  • 325.38€ / 150µL
Expédition estimée: 03-10-2022
pour voir le stock disponible 
Description
Description
The PREX2 antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human PREX2. This antibody recognizes human antigen. The PREX2 antibody has been validated for the following applications: IF, IHC, ELISA analysis.
Spécification
Spécification
PREX2
Polyclonal
Unconjugated
150 μL
80243
IgG
ELISA, Immunofluorescence, Immunohistochemistry
360 μg/mL