Learn More
Epredia™ Lab Vision™ Protein Gene Product 9.5 (PGP9.5) Ab-1, Mouse Monoclonal Antibody
Provide accurate, reproducible results with the Epredia™ Protein Gene Product 9.5 (PGP9.5) Ab-1, Mouse Monoclonal Antibody.
Marque: Epredia™ MS-1760-R7
Description
Protein gene product 9.5 is a ubiquitin C-terminal hydroxylase found in neurons and nerve fibres of central and peripheral nervous system; many neuroendocrinal cells, segments of renal tubules, spermatogonia, leydig cells of testis; and ova. It is present in cellular inclusions and can be a useful as a neuronal marker and in the studies of neurodegenerative disorders.Host Species: Mouse
Clone: 10A1
Isotype: IgG2b
Species Reactivity: Human. Others not known.
Epitope: Not determined
Immunogen: Recombinant protein corresponding to full-length human protein gene product 9.5 molecule
Molecular Weight: 25-27kDa
Positive Control: Small intestine
Cellular Localization: Cytoplasmic and nuclear
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Spécification
| Protein Gene Product 9.5 (PGP9.5) Ab-1 | |
| Monoclonal | |
| Unconjugated | |
| Recombinant protein corresponding to full length human protein gene product 95 molecule | |
| IVD | |
| Primary | |
| Prediluted |
| Immunohistochemistry (Paraffin) | |
| 10A1 | |
| Mouse | |
| 7 mL | |
| Neuroscience | |
| Human | |
| IgG2b |
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.