Learn More
Epredia™ Lab Vision™ Protein Gene Product 9.5 (PGP9.5) Ab-1, Mouse Monoclonal Antibody
Provide accurate, reproducible results with the Epredia™ Protein Gene Product 9.5 (PGP9.5) Ab-1, Mouse Monoclonal Antibody.
94.35€ - 403.92€
Spécification
| Antigène | Protein Gene Product 9.5 (PGP9.5) Ab-1 |
|---|---|
| Clone | 10A1 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Conjugué | Unconjugated |
| Code produit | Marque | Quantité | Forme | Prix | Quantité et disponibilité | ||||
|---|---|---|---|---|---|---|---|---|---|
| Code produit | Marque | Quantité | Forme | Prix | Quantité et disponibilité | ||||
|
12690117
|
Epredia™
MS-1760-S0 |
100 μL | Concentrated | ||||||
|
12600127
|
Epredia™
MS-1760-S1 |
500 μL | Concentrated | ||||||
|
12680117
|
Epredia™
MS-1760-S |
1 mL | Concentrated | ||||||
|
12670117
|
Epredia™
MS-1760-R7 |
7 mL | Prediluted | ||||||
Description
Protein gene product 9.5 is a ubiquitin C-terminal hydroxylase found in neurons and nerve fibres of central and peripheral nervous system; many neuroendocrinal cells, segments of renal tubules, spermatogonia, leydig cells of testis; and ova. It is present in cellular inclusions and can be a useful as a neuronal marker and in the studies of neurodegenerative disorders.Host Species: Mouse
Clone: 10A1
Isotype: IgG2b
Species Reactivity: Human. Others not known.
Epitope: Not determined
Immunogen: Recombinant protein corresponding to full-length human protein gene product 9.5 molecule
Molecular Weight: 25-27kDa
Positive Control: Small intestine
Cellular Localization: Cytoplasmic and nuclear
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
1. Wilkinson K D, et al. (1989) Science, 246:670-675.
2. Wilson P O G, et al. (1988) Br J Exp Pathol, 69:91-104.
Spécification
| Protein Gene Product 9.5 (PGP9.5) Ab-1 | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| IVD | |
| Human | |
| IgG2b |
| 10A1 | |
| Monoclonal | |
| Mouse | |
| Neuroscience | |
| Recombinant protein corresponding to full length human protein gene product 95 molecule | |
| Primary |
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.