missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRR5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 17948-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
MARK1 (MAP/microtubule affinity-regulating kinase 1) belongs to the MARK family of serine/threonine kinases. MARK family protein kinases phosphorylate microtubule-associated proteins and may play a role in cytoskeletal stability. MARK2 refers to MAP/microtubule affinity-regulating kinase 2 isoform a [Homo sapiens]. EMK (ELKL Motif Kinase) is a small family of ser/thr protein kinases involved in the control of cell polarity, microtubule stability and cancer. Several cDNA clones have been isolated that encoded two isoforms of the human ser/thr protein kinase EMK1 called MARK2. MARK3 was originally identified as a marker that was induced by treatment with DNA damaging agents, and loss of MARK3 was found with carcinogenesis in the pancreas. MARK4 contains an N terminal serine/threonine kinase domain, a central ubiquitin associated domain, and a C terminal KA1 associated kinase domain. RT PCR analysis detects upregulated expression of the gene for MARK4 in nearly all clinical hepatocellular carcinoma cells.Spécification
PRR5 | |
Polyclonal | |
Unconjugated | |
PRR5 | |
FLJ20185k, PP610, proline rich 5 (renal), Proline rich protein 5, Protein observed with Rictor 1, Protor 1, PROTOR1, PRR5 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
109270, 315189, 55615 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.15 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P85299, Q5FVG6, Q812A5 | |
Prr5 | |
PRR5 Fusion Protein Ag12424 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |