missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSMC4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 11389-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
GTPase-activating protein for the ADP ribosylation factor family.Spécification
PSMC4 | |
Polyclonal | |
Unconjugated | |
PSMC4 | |
MB67 interacting protein, MIP224, PSMC4, S6, Tat binding protein 7, TBP 7, TBP7 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
117262, 23996, 5704 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot, Immunohistochemistry (Paraffin) | |
0.29 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P43686, P54775, Q63570 | |
PSMC4 | |
PSMC4 Fusion Protein Ag1943 | |
20 μL | |
Primary | |
Rat, Human, Mouse | |
Antibody | |
IgG |