missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PTPN4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 11131-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
The protein encoded by this gene is a member of the protein tyrosine phosphatase family. PTPs are known to be signaling molecules that regulate a variety of cellular processes including cell growth, differentiation, mitotic cycle, and oncogenic transformation. This protein contains a C-terminal PTP domain and an N-terminal domain homologous to the band 4.1 superfamily of cytoskeletal-associated proteins. This PTP has been shown to interact with glutamate receptor delta 2 and epsilon subunits, and is thought to play a role in signaling downstream of the glutamate receptors through tyrosine dephosphorylation.Spécification
PTPN4 | |
Polyclonal | |
Unconjugated | |
PTPN4 | |
MEG, PTPase MEG1, PTPMEG, PTPMEG1, PTPN4 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
19258, 246116, 5775 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.37 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P29074, Q9WU22 | |
PTPN4 | |
PTPN4 Fusion Protein Ag1585 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |