missing translation for 'onlineSavingsMsg'
Learn More

RAB5A Mouse anti-Human, Porcine, Clone: 1B6A5, Proteintech

Mouse Monoclonal Antibody

Marque:  Proteintech 66339-1-IG-150UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16828125

  • 325.38€ / 150µL
Expédition estimée: 03-10-2022
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

RAB5 are among the small GTPases Rab are key regulators of intracellular membrane trafficking, from the formation of transport vesicles to their fusion with membranes. Rabs cycle between an inactive GDP-bound form and an active GTP-bound form that is able to recruit to membranes different sets of downstream effectors directly responsible for vesicle formation, movement, tethering and fusion. RAB5A is required for the fusion of plasma membranes and early endosomes. Contributes to the regulation of filopodia extension. Required for the exosomal release of SDCBP, CD63, PDCD6IP and syndecan. Regulates maturation of apoptotic cell-containing phagosomes, probably downstream of DYN2 and PIK3C3.
Spécification

Spécification

RAB5A
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P20339, Q06AU6
RAB5A
RAB5A Fusion Protein Ag16781
150 μL
Primary
Porcine, Human
Antibody
IgG2b
Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry
1B6A5
Unconjugated
RAB5A
RAB5, RAB5A, Ras related protein Rab 5A
Mouse
Protein A
RUO
100144499, 5868
-20°C
Liquid
Vidéos
FDS
Documents
Certificats
Promotions

Promotions