Learn More
MYC tag, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of MYC tag.
Marque: Abnova PAB12716.100ug
Description
The protein encoded by this gene is a multifunctional, nuclear phosphoprotein that plays a role in cell cycle progression, apoptosis and cellular transformation. It functions as a transcription factor that regulates transcription of specific target genes. Mutations, overexpression, rearrangement and translocation of this gene have been associated with a variety of hematopoietic tumors, leukemias and lymphomas, including Burkitt lymphoma. There is evidence to show that alternative translation initiations from an upstream, in-frame non-AUG (CUG) and a downstream AUG start site result in the production of two isoforms with distinct N-termini. The synthesis of non-AUG initiated protein is suppressed in Burkitt's lymphomas, suggesting its importance in the normal function of this gene. [provided by RefSeq]
Sequence: EQKLISEEDLSpécification
MYC tag | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of MYC tag. | |
In TBS, pH 7.2 (BSA, 10% Proclin300) | |
bHLHe39/c-Myc | |
Rabbit | |
100 μg | |
Primary | |
This antibody recognizes MYC tagged fusion proteins. | |
Store at 4°C. For long term storage store at -20°C or -80°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG |
ELISA, Immunoprecipitation, Western Blot | |
Unconjugated | |
Western Blot (0.1-1 ug/mL) ELISA (0.01-0.1 ug/mL) Immunoprecipitation (2-5 ug/mL) The optimal working dilution should be determined by the end user. | |
MYC | |
MYC | |
A synthetic peptide corresponding to amino acids 408-452 of human MYC tag. | |
RUO | |
4609 | |
Human | |
Liquid |
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
For Research Use Only