missing translation for 'onlineSavingsMsg'
Learn More

RAC2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 20573-1-AP-20UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16830135

  • 121.38€ / 20µL
Expédition estimée: 06-10-2022
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

The antibody is specific to RAC2.

RAC, a small GTPase in the ras family, regulates the formation of membrane ruffles, lamellipodia and filopodia. It is involved in cytoskeletal actin organization and transformation. The p21 activated kinases (PAKs) are direct targets of active Rac and Cdc42 which can induce the assembly of polarized cytoskeletal structures when expressed in fibroblasts. RAC1 and RAC2 are also essential components of NADPH oxidase, the enzyme responsible for generating free radicals.
Spécification

Spécification

RAC2
Polyclonal
Unconjugated
RAC2
EN 7, Gx, HSPC022, p21 Rac2, RAC2, Small G protein
Rabbit
Antigen Affinity Chromatography
RUO
19354, 5880
-20°C
Liquid
Western Blot
0.5 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P15153, Q05144
RAC2
Peptide
20 μL
Primary
Mouse, Human
Antibody
IgG
Vidéos
FDS
Documents
Certificats
Promotions

Promotions