Learn More
RAN Mouse anti-Human, Unconjugated, Clone: 1B11E1, Proteintech
Mouse Monoclonal Antibody
Marque:  Proteintech 67500-1-IG-150UL
Informations supplémentaires : Poids : 0.00010kg
 Afficher plus de versions de ce produit
Code produit. 16830166
  • 325.38€ / 150µL
Expédition estimée: 03-10-2022
pour voir le stock disponible 
Description
Description
The RAN antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human RAN. This antibody recognizes human, mouse, rat antigen. The RAN antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Spécification
Spécification
RAN
Monoclonal
1000 μg/mL
Mouse
Primary
Mouse, Rat, Human
Immunofluorescence, Immunohistochemistry, Western Blot
1B11E1
Unconjugated
150 μL
5901
IgG2b