missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Raptor Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Marque: Proteintech 68286-1-IG-20UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
The Raptor antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human Raptor. This antibody recognizes human antigen. The Raptor antibody has been validated for the following applications: WB, IP, ELISA analysis.Spécification
| Raptor | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Protein A purification | |
| RUO | |
| 57521 | |
| Store at -20°C. Stable for one year after shipment. | |
| Monoclonal Antibody | |
| IgG2b |
| Western Blot, Immunoprecipitation, ELISA | |
| 1B8A12 | |
| Unconjugated | |
| Recombinant Protein | |
| 20 μL | |
| Primary | |
| Human | |
| 130-150 kDa | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction