missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RBBP9 Mouse anti-Human, Clone: 3D5E11, Proteintech
Mouse Monoclonal Antibody
Marque: Proteintech 66015-2-IG-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
RBBP9 may play a role in the transformation process due to its capacity to confer resistance to the growth-inhibitory effects of TGF-beta1 through interaction with retinoblastoma and the subsequent displacement of E2F-1.The protein encoded by this gene is a retinoblastoma binding protein that may play a role in the regulation of cell proliferation and differentiation. Two alternatively spliced transcript variants of this gene with identical predicted protein products have been reported, one of which is a nonsense-mediated decay candidate.Spécification
RBBP9 | |
Monoclonal | |
0.81 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O75884 | |
RBBP9 | |
RBBP9 Fusion Protein Ag17864 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG2b |
Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry | |
3D5E11 | |
Unconjugated | |
RBBP9 | |
BOG, RBBP9 | |
Mouse | |
Protein A | |
RUO | |
10741 | |
-20°C | |
Liquid |