missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RBMX2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 17994-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Spécification
RBMX2 | |
Polyclonal | |
Unconjugated | |
RBMX2 | |
CGI 79, RBMX2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
209003, 367930, 51634 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.15 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
B0BN49, Q8R0F5, Q9Y388 | |
Rbmx2 | |
RBMX2 Fusion Protein Ag12439 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |