missing translation for 'onlineSavingsMsg'
Learn More
Learn More
REG4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 12268-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
Reg IV is part of the regenerating gene family within the C-type lectin superfamily. This family is involved in liver, pancreatic, gastric and intestinal cell proliferation and differentiation. Reg IV is a 158-amino acid secretory protein implicated in cell regeneration and/or survival with a definite growth stimulating effect on pancreatic beta cells. It is highly expressed in colorectal, gastric, prostate and other types of cancer. Reg IV-positive tumor cells display different phenotypes including mucus-secreting, enterocyte-like, and undifferentiated.Spécification
REG4 | |
Polyclonal | |
Unconjugated | |
REG4 | |
GISP, REG 4, REG IV, REG like protein, REG4, RELP | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
445583, 67709, 83998 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.29 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q68AX7, Q9BYZ8, Q9D8G5 | |
Reg4 | |
REG4 Fusion Protein Ag2934 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |