missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RILP Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 13574-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
The Rab interacting lysosomal protein (RILP) is lysosomal protein that interacts with RAB7, a small GTPase that controls transport to endocytic degradative compartments. It is thought that RILP is a downstream effector of RAB7 and both proteins act together in the regulation of late endocytic traffic. Overexpression of RILP caused enlarged lysosomes that are more centrally located in the cell suggesting that RILP may also be involved in the regulation of lysosomal morphology. Other evidence suggests that RILP may play a role in the biogenesis of multivesicular bodies.Spécification
RILP | |
Polyclonal | |
Unconjugated | |
RILP | |
PP10141, RILP | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
83547 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.58 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q96NA2 | |
RILP | |
RILP Fusion Protein Ag4472 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |