missing translation for 'onlineSavingsMsg'
Learn More

RPAIN Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 15220-1-AP-20UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16834314

  • 121.38€ / 20µL
Expédition estimée: 10-10-2022
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Replication protein A (RPA) is a single-stranded-DNA binding protein involved in numerous eukaryotic DNA processes including replication, repair and recombination. RPA interacting protein (RPA IP) has been identified as an adapter protein that is involved in RPA nuclear import instead of the prototypical importin proteins that normally mediate nuclear import. Multiple isoforms of RPA IP are known to exist, with the longest isoform localized to the cytoplasm. Isoform 2 is sumoylated and is located in the PML nuclear body within the nucleus. It has been suggested that this isoform mediates the localization of the RPA complex into the PML nuclear body, thereby participating in RPA function in DNA metabolism.
Spécification

Spécification

RPAIN
Polyclonal
Unconjugated
RPAIN
HRIP, RIP, RPA interacting protein, RPAIN
Rabbit
Antigen Affinity Chromatography
RUO
69723, 84268
-20°C
Liquid
Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin)
0.2 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q86UA6, Q9CWY9
Rpain
RPAIN Fusion Protein Ag7392
20 μL
Primary
Human, Mouse
Antibody
IgG
Vidéos
FDS
Documents
Certificats
Promotions

Promotions