missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPIA Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 13010-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
Defects in RPIA are the cause of ribose 5-phosphate isomerase deficiency. The exact function of RPIA remains unknown.Spécification
RPIA | |
Polyclonal | |
Unconjugated | |
RPIA | |
Phosphoriboisomerase, Ribose 5 phosphate isomerase, ribose 5 phosphate isomerase A, RPI, RPIA | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
19895, 22934, 362383 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.21 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P47968, P49247 | |
Rpia | |
RPIA Fusion Protein Ag3699 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |