Learn More
RPL11 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque:  Proteintech 16277-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Code produit. 16835864
  • 140.00€ / 20µL
Expédition estimée: 15-04-2024
Description
The RPL11 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human RPL11. This antibody recognizes human, mouse, rat antigen. The RPL11 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Spécification
Spécification
RPL11
Polyclonal
Unconjugated
20 μL
6135
IgG
Western Blot, Immunoprecipitation, Immunohistochemistry, Immunofluorescence, ELISA
207 μg/mL
Rabbit
Primary
Human, Rat, Mouse