Learn More
RPL11 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 16277-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
The RPL11 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human RPL11. This antibody recognizes human, mouse, rat antigen. The RPL11 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Spécification
RPL11 | |
Polyclonal | |
Unconjugated | |
20 μL | |
6135 | |
IgG |
Western Blot, Immunoprecipitation, Immunohistochemistry, Immunofluorescence, ELISA | |
207 μg/mL | |
Rabbit | |
Primary | |
Human, Rat, Mouse |