missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPL23A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 16386-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
The function of this protein remains unknown.Spécification
RPL23A | |
Polyclonal | |
Unconjugated | |
RPL23A | |
60S ribosomal protein L23a, MDA20, ribosomal protein L23a, RPL23A | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
268449, 360572, 6147 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P62750, P62751, P62752 | |
Rpl23a | |
RPL23A Fusion Protein Ag9424 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |