missing translation for 'onlineSavingsMsg'
Learn More

RPLP0 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 51019-2-AP-150UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16836285

  • 325.38€ / 150µL
Expédition estimée: 03-10-2022
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

The protein encoded by this gene contains RNA recognition motif found in a variety of RNA binding proteins, including various hnRNP proteins, proteins implicated in regulation of alternative splicing, and protein components of snRNPs. In vitro studies showed that the rat homolog bound to RNA homopolymers, with a preference for G and U polyribonucleotides. This gene is part of a gene cluster on chromosome Xp11.23, and its 3′ end lies within 20 kb upstream of UBE1. Two transcript variants encoding different isoforms have been identified for this gene.
Spécification

Spécification

RPLP0
Polyclonal
Unconjugated
RPLP0
60S ribosomal protein L10E, L10E, P0, PRLP0, ribosomal protein, large, P0, RPLP0, RPP0
Rabbit
Antigen Affinity Chromatography
RUO
11837, 6175, 64205
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin)
0.13 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P05388, P14869, P19945
RPLP0
Peptide
150 μL
Primary
Rat, Mouse, Human
Antibody
IgG
Vidéos
FDS
Documents
Certificats
Promotions

Promotions