missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPP38 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 12874-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
Component of ribonuclease P, a protein complex that generates mature tRNA molecules by cleaving their 5′-ends. RPP38 may associate transiently with RNase P RNA as a factor involved in the transport of H1 RNA to the putative site of its assembly in the cell, the nucleolus.Spécification
RPP38 | |
Polyclonal | |
Unconjugated | |
RPP38 | |
RNaseP protein p38, RP11 455B2.5, RPP38 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10557, 227522, 291317 | |
-20°C | |
Liquid |
Western Blot | |
0.61 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P78345, Q80UU2 | |
Rpp38 | |
RPP38 Fusion Protein Ag3552 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |