missing translation for 'onlineSavingsMsg'
Learn More

RPP38 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 12874-1-AP-20UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16830824

  • 140.00€ / 20µL
Expédition estimée: 31-05-2024
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Component of ribonuclease P, a protein complex that generates mature tRNA molecules by cleaving their 5′-ends. RPP38 may associate transiently with RNase P RNA as a factor involved in the transport of H1 RNA to the putative site of its assembly in the cell, the nucleolus.
Spécification

Spécification

RPP38
Polyclonal
Unconjugated
RPP38
RNaseP protein p38, RP11 455B2.5, RPP38
Rabbit
Antigen Affinity Chromatography
RUO
10557, 227522, 291317
-20°C
Liquid
Western Blot
0.61 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P78345, Q80UU2
Rpp38
RPP38 Fusion Protein Ag3552
20 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions