missing translation for 'onlineSavingsMsg'
Learn More

RPS3A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 14123-1-AP-20UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16832624

  • 121.38€ / 20µL
Expédition estimée: 28-09-2022
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 40S subunit. The protein belongs to the S3AE family of ribosomal proteins. It is located in the cytoplasm. Disruption of the gene encoding rat ribosomal protein S3a, also named v-fos transformation effector protein, in v-fos-transformed rat cells results in reversion of the transformed phenotype. This gene is co-transcribed with the U73A and U73B small nucleolar RNA genes, which are located in its fourth and third introns, respectively. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome. Alternatively spliced transcript variants have been found for this gene.
Spécification

Spécification

RPS3A
Polyclonal
Unconjugated
RPS3A
40S ribosomal protein S3a, Fte 1, FTE1, MFTL, ribosomal protein S3A, RPS3A
Rabbit
Antigen Affinity Chromatography
RUO
20091, 29288, 6189
-20°C
Liquid
Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Flow Cytometry, Immunocytochemistry
0.27 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P49242, P61247, P97351
RPS3A
RPS3A Fusion Protein Ag5273
20 μL
Primary
Mouse, Human, Rat
Antibody
IgG
Vidéos
FDS
Documents
Certificats
Promotions

Promotions