missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPTOR Antibody (SR1170), Novus Biologicals™
Rabbit Monoclonal Antibody
Marque: Novus Biologicals NBP3-21541-50ul
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
RPTOR Monoclonal antibody specifically detects RPTOR in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunoprecipitation
Spécification
| RPTOR | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunoprecipitation | |
| SR1170 | |
| Western Blot 1:500-1:1000, Flow Cytometry 1:30, Immunohistochemistry 1:50-1:100, Immunocytochemistry/Immunofluorescence 1:50-1:100, Immunoprecipitation 1:30 | |
| KIAA1303p150 target of rapamycin (TOR)-scaffold protein containing WD-repeats, KOG1, Mip1, p150 target of rapamycin (TOR)-scaffold protein, Raptor, regulatory associated protein of MTOR, complex 1, regulatory-associated protein of mTOR | |
| A synthesized peptide derived from human RPTOR (Uniprot #: Q8N122) | |
| 50 μg | |
| Autophagy, Cancer, Hypoxia, mTOR Pathway, Signal Transduction | |
| 57521 | |
| Store at -20 degrees C. Avoid freeze/thaw cycles. | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu