missing translation for 'onlineSavingsMsg'
Learn More

RYR2 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 27587-1-AP-150UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16835715

  • 399.00€ / 150µL
Expédition estimée: 15-04-2024
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Dihydropyridine receptor (DHPR) is a surface membrane protein critical for the excitation-contraction coupling of striated muscle. DHPR and the sarcoplasmic reticulum ryanodine receptor (RyR) are two key components of the intracellular junctions, where depolarization of the surface membrane is converted into the release of Ca2+ from internal stores. The alpha1-subunit of the DHPR contains a cytoplasmic loop which is thought to be involved in the interactions with RyR. Phosphorylation of the DHPR alpha1-subunit is also thought to play a role in the functional interaction of DHPR and RyR. Mutation in DHPR alpha1 results in excitation-contraction uncoupling, leading to muscular dysgenesis, a complete inactivity in developing skeletal muscles. Cells that do not express RyR also lack excitation-contraction coupling and exhibit a severalfold reduction in Ca2+ current density.
Spécification

Spécification

RYR2
Polyclonal
Unconjugated
RYR2
ARVC2, ARVD2, ryanodine receptor 2 (cardiac), RYR2, VTSIP
Rabbit
Antigen Affinity Chromatography
RUO
6262
-20°C
Liquid
Immunohistochemistry (Paraffin)
0.43 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q92736
RYR2
RYR2 Fusion Protein Ag26296
150 μL
Primary
Human
Antibody
IgG
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions