Learn More
Sam50 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 28679-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
The Sam50 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human Sam50. This antibody recognizes human, mouse, rat antigen. The Sam50 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Spécification
Sam50 | |
Polyclonal | |
Unconjugated | |
150 μL | |
25813 | |
IgG |
Western Blot, Immunohistochemistry, Immunofluorescence | |
400 μg/mL | |
Rabbit | |
Primary | |
Human, Rat, Mouse |