Learn More
SARS-COV-2 NSP7 Rabbit, anti-Virus, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 29263-1-AP-20UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
The SARS-COV-2 NSP7 antibody from Proteintech is a rabbit polyclonal antibody to a recombinant protein of sars-cov-2 SARS-COV-2 NSP7. This antibody recognizes virus antigen. The SARS-COV-2 NSP7 antibody has been validated for the following applications: ELISA analysis.
Spécification
| SARS-COV-2 NSP7 | |
| Polyclonal | |
| Unconjugated | |
| 20 μL | |
| 43740578 | |
| Liquid |
| ELISA | |
| 350 μg/mL | |
| Rabbit | |
| Primary | |
| Virus |