missing translation for 'onlineSavingsMsg'
Learn More

SCCPDH Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 24941-1-AP-150UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16833465

  • 325.38€ / 150µL
Expédition estimée: 06-10-2022
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

SCCPDH (Probable saccharopine dehydrogenase) is a 429 amino acid protein that belongs to the saccharopine dehydrogenase family. The SCCPDH gene is conserved in chimpanzee, dog, cow, mouse, rat, chicken, fruit fly, mosquito and C.elegans, and maps to human chromosome 1q44. Chromosome 1 is the largest human chromosome spanning about 260 million base pairs and making up 8% of the human genome. There are about 3,000 genes on chromosome 1, and considering the great number of genes there are also a large number of diseases associated with chromosome 1. Stickler syndrome, Parkinsons, Gaucher disease and Usher syndrome are also associated with chromosome 1. A breakpoint has been identified in 1q which disrupts the DISC1 gene and is linked to schizophrenia. Aberrations in chromosome 1 are found in a variety of cancers including head and neck cancer, maligt melanoma and multiple myeloma.
Spécification

Spécification

SCCPDH
Polyclonal
Unconjugated
SCCPDH
CGI 49, NET11, RP11 439E19.2, SCCPDH
Rabbit
Antigen Affinity Chromatography
RUO
109232, 51097
-20°C
Liquid
Western Blot, Immunoprecipitation, Immunofluorescence, Immunocytochemistry
0.35 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q8NBX0, Q8R127
SCCPDH
SCCPDH Fusion Protein Ag11551
150 μL
Primary
Mouse, Human
Antibody
IgG
Vidéos
FDS
Documents
Certificats
Promotions

Promotions