missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SDR42E1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 17347-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Spécification
SDR42E1 | |
Polyclonal | |
Unconjugated | |
SDR42E1 | |
HSPC105, SDR42E1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
307897, 74032, 93517 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.21 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8WUS8, Q9D665 | |
Sdr42e1 | |
SDR42E1 Fusion Protein Ag11391 | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |