missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SDS3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Marque: Novus Biologicals NBP2-48751
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
SDS3 Polyclonal antibody specifically detects SDS3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Spécification
| SDS3 | |
| Polyclonal | |
| Western Blot 0.04-0.4 μg/mL, Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| 45 kDa Sin3-associated polypeptide, FLJ00052, SAP45suppressor of defective silencing 3 homolog (SDS3, S. cerevisiae), SDS3MGC104711, sin3 histone deacetylase corepressor complex component SDS3, sin3A-associated protein, 45kDa, suppressor of defective silencing 3 homolog (S. cerevisiae), Suppressor of defective silencing 3 protein homolog | |
| This antibody was developed against a recombinant protein corresponding to amino acids: KAAVKEFEDKKVELKENLIAELEEKKKMIENEKLTMELTGDSMEVKPIMTRKLRRRPNDPVPIPDKRRKPAPAQLNYLLTDEQIMEDLRTLNKLKSPKRPA | |
| 0.1 mL | |
| DNA Repair | |
| 64426 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2), 40% Glycerol | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu