missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SF3B1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 27684-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
This gene belongs to a group of vacuolar protein sorting (VPS) genes. The encoded protein is a component of a large multimeric complex, termed the retromer complex, involved in retrograde transport of proteins from endosomes to the trans-Golgi network. The close structural similarity between the yeast and human proteins that make up this complex suggests a similarity in function. Expression studies in yeast and mammalian cells indicate that this protein interacts directly with VPS35, which serves as the core of the retromer complex.Spécification
SF3B1 | |
Polyclonal | |
Unconjugated | |
SF3B1 | |
PRP10, PRPF10, SAP 155, SAP155, SF3B1, SF3b155, Splicing factor 3B subunit 1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
23451 | |
-20°C | |
Liquid |
Western Blot | |
0.47 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O75533 | |
SF3B1 | |
SF3B1 Fusion Protein Ag26689 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |