missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SHD Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 20814-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Spécification
SHD | |
Polyclonal | |
Unconjugated | |
SHD | |
SHD | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
20420, 56961 | |
-20°C | |
Liquid |
Immunofluorescence, Immunocytochemistry, Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O88834, Q96IW2 | |
SHD | |
SHD Fusion Protein Ag14794 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |