missing translation for 'onlineSavingsMsg'
Learn More

Siglec-1/CD169 Antibody (3D6.112 (recombinant version)) - Chimeric - Azide and BSA Free, Novus Biologicals™

Rabbit Monoclonal Antibody

282.00€ - 928.00€

Specifications

Antigène Siglec-1/CD169
Clone 3D6.112 (recombinant version)
Concentration 1 mg/mL
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantité Price Quantity & Availability  
Product Code Brand Quantité Price Quantity & Availability  
18617606
missing translation for 'viewDocuments'
Novus Biologicals
NBP3-08989-0.025mg
0.025 mg
282.00€
25µg
View alternative products
missing translation for 'toSaveUpto' 
»
Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!
18644027
missing translation for 'viewDocuments'
Novus Biologicals
NBP3-08989-0.1mg
0.1 mg
928.00€
0.10mg
View alternative products
missing translation for 'toSaveUpto' 
»
Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!
Description

Description

Siglec-1/CD169 Monoclonal antibody specifically detects Siglec-1/CD169 in Mouse samples. It is validated for Western Blot, Immunohistochemistry
Specifications

Specifications

Siglec-1/CD169
1 mg/mL
Monoclonal
Purified
RUO
Mouse
CD169, FLJ00051, sialic acid binding Ig-like lectin 1, sialoadhesin, sialoadhesin
Spleen cells from AO rats immunised with thioglycollate-elicted murine peritoneal macrophages (TPM) that had been cutured in mouse serum to induce sheep erythrocyte receptor (SER).
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
3D6.112 (recombinant version)
Western Blot, Immunohistochemistry
Unconjugated
Rabbit
Adaptive Immunity, Immunology
PBS
6614
IgG κ
Protein A purified
Videos
SDS
Documents

Documents

Certificates
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Siglec-1/CD169 Antibody (3D6.112 (recombinant version)) - Chimeric - Azide and BSA Free, Novus Biologicals™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.